Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,6,6-Tetramethyl-3,5-heptanedione, 98%

Catalog No. AAA1511814
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1511814 25 g
AAA1511806 5 g
AAA1511822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1511814 Supplier Thermo Scientific Chemicals Supplier No. A1511814
Add to Cart

CAS: 1118-71-4 | C11H20O2 | 184.279 g/mol

2,2,6,6-Tetramethyl-3,5-heptanedione is used in the synthesis of α-aryl-β-diketones1 and dicyanamidobenzene-bridge diruthenium complex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,6,6-Tetramethyl-3,5-heptanedione is used in the synthesis of α-aryl-β-diketones1 and dicyanamidobenzene-bridge diruthenium complex.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1118-71-4
Molecular Formula C11H20O2
Molecular Weight (g/mol) 184.279
MDL Number MFCD00008848
InChI Key YRAJNWYBUCUFBD-UHFFFAOYSA-N
Synonym dipivaloylmethane, 2,2,6,6-tetramethyl-3,5-heptanedione, 3,5-heptanedione, 2,2,6,6-tetramethyl, unii-r8ui909hoy, tmhd, 2,2,6,6-tetramethyl-3,5-heptanedione dipivaloylmethane, r8ui909hoy, 2,2,6,6-tetramethyl-heptane-3,5-dione, pubchem12497
PubChem CID 70700
IUPAC Name 2,2,6,6-tetramethylheptane-3,5-dione
SMILES CC(C)(C)C(=O)CC(=O)C(C)(C)C

Specifications

Density 0.889
Boiling Point 72°C to 73°C (6 mmHg)
Flash Point 67°C (152°F)
Refractive Index 1.459
Quantity 25 g
Beilstein 1447269
Solubility Information Difficult to mix.
Formula Weight 184.28
Percent Purity 98%
Chemical Name or Material 2,2,6,6-Tetramethyl-3,5-heptanedione
Hazard Category H227-H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H227
Harmful if swallowed.
Combustible liquid.
Precautionary Statement P210-P233-P240-P243-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P374-P501c
EINECSNumber 214-268-3
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.