Learn More
Sigma Organic Chemistry 2 2 6 6-Tetramethyl-3 6 | 1G | 1142363-56-1 | MFCD11044439
Supplier: Sigma Organic Chemistry BML002181G

2 2 6 6-Tetramethyl-3 6 , 1G
About this Item:
CAS #: 1142363-56-1
MDL #: MFCD11044439
Molecular Weight: 266.18
UNSPSC Code: 12352200
Chemical Formula: C15H27BO3
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.