Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,6,6-Tetramethyl-4-piperidone 98.0+%, TCI America™
SDP

Catalog No. T142425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T142425G 25 g
T1424100G 100 g
T1424500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T142425G Supplier TCI America Supplier No. T142425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 826-36-8
Molecular Formula C9H17NO
Molecular Weight (g/mol) 155.241
MDL Number MFCD00149393
InChI Key JWUXJYZVKZKLTJ-UHFFFAOYSA-N
Synonym triacetonamine, 2,2,6,6-tetramethyl-4-piperidone, triacetone amine, triacetonamin, vincubina, vincubine, 2,2,6,6-tetramethyl-4-oxopiperidine, tempidon, 2,2,6,6-tetramethyl-4-piperidinone, trojacetonoaminy
PubChem CID 13220
IUPAC Name 2,2,6,6-tetramethylpiperidin-4-one
SMILES CC1(CC(=O)CC(N1)(C)C)C

Specifications

Melting Point 38°C
Boiling Point 105°C
Quantity 25 g
Formula Weight 155.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2,6,6-Tetramethyl-4-piperidone
EINECSNumber (5)-0777
RTECSNumber TO0127900
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.