Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2',6-Tribromoindophenol Sodium Salt, TCI America™
SDP

Catalog No. T03531G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T03531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T03531G Supplier TCI America Supplier No. T03531G

Chemical Identifiers

CAS 5418-30-4
Molecular Formula C12H5Br3NNaO2
Molecular Weight (g/mol) 457.879
MDL Number MFCD00059569
InChI Key AOFQQAPWDVFFEH-UHFFFAOYSA-M
Synonym 2,6-Dibromo-2′C-bromo-indophenol Sodium Salt, 2,6-Dibromophenol-indo-m-bromophenol Sodium Salt
PubChem CID 44630496
IUPAC Name sodium;3-bromo-4-[(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)amino]phenolate
SMILES C1=CC(=C(C=C1[O-])Br)N=C2C=C(C(=O)C(=C2)Br)Br.[Na+]

Specifications

Quantity 1 g
Formula Weight 457.88
Chemical Name or Material 2,2′,6-Tribromoindophenol Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.