Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Bipyridine, ACS, 98%

Catalog No. AA3056918
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3056918 50 g
AA3056909 10 g
AA3056930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3056918 Supplier Thermo Scientific Chemicals Supplier No. 03056918
Only null left
Add to Cart

CAS: 366-18-7 | C10H8N2 | 156.19 g/mol

2,2'-bipyridine acts as a bidentate chelating ligand which form complexes with transition metal ions and shows antifungal, antibacterial and antiviral activity. It is used for the colorimetric determination of iron as well as oxidation reduction indicators to confirm the presence of ferrous ion in soils. It forms complexes with ruthenium and platinum, exhibit intense luminescence, which may have practical applications. Copper(I) bipyridine complexes involved in the oxidation of alcohols under aerobic conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 366-18-7
Molecular Formula C10H8N2
Molecular Weight (g/mol) 156.19
MDL Number MFCD00006212
InChI Key ROFVEXUMMXZLPA-UHFFFAOYSA-N
Synonym 2,2'-bipyridine, 2,2'-dipyridyl, 2,2'-bipyridyl, bipyridine, 2,2'-dipyridine, 2-2-pyridyl pyridine, 2,2'-bipyridin, alpha,alpha'-dipyridyl, alpha,alpha'-bipyridyl, 2,2-bipyridyl
PubChem CID 1474
ChEBI CHEBI:30351
SMILES C1=CC=C(N=C1)C1=CC=CC=N1

Specifications

Melting Point 70°C
Boiling Point 272°C to 273°C
Flash Point 121°C (249°F)
Quantity 50 g
UN Number UN2811
Beilstein 113089
Merck Index 14,3347
Solubility Information Soluble in organic solvents and slightly soluble in water.
IUPAC Name 2,2'-bipyridine
Formula Weight 156.19
Percent Purity 98%
Grade ACS
Physical Form Crystalline
Chemical Name or Material 2, 2'-Bipyridyl
Show More Show Less
Hazard Category H301-H312-H500
Hazard Statement GHS H Statement
H301-H312
Precautionary Statement P264b-P270-P280-P301+P310-P302+P352-P312-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 206-674-4
RTECSNumber DW1750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.