CAS RN 366-18-7
2,2'-Dipyridyl, 99+%
CAS: 366-18-7 Molecular Formula: C10H8N2 Molecular Weight (g/mol): 156.19 MDL Number: MFCD00006212 InChI Key: ROFVEXUMMXZLPA-UHFFFAOYSA-N Synonym: 2,2'-bipyridine,2,2'-dipyridyl,2,2'-bipyridyl,bipyridine,2,2'-dipyridine,2-2-pyridyl pyridine,2,2'-bipyridin,alpha,alpha'-dipyridyl,alpha,alpha'-bipyridyl,2,2-bipyridyl PubChem CID: 1474 ChEBI: CHEBI:30351 IUPAC Name: 2-pyridin-2-ylpyridine SMILES: C1=CC=C(N=C1)C1=CC=CC=N1
2,2'-Bipyridine, 99+%
CAS: 366-18-7 Molecular Formula: C10H8N2 Molecular Weight (g/mol): 156.19 MDL Number: MFCD00006212 InChI Key: ROFVEXUMMXZLPA-UHFFFAOYSA-N Synonym: 2,2'-bipyridine,2,2'-dipyridyl,2,2'-bipyridyl,bipyridine,2,2'-dipyridine,2-2-pyridyl pyridine,2,2'-bipyridin,alpha,alpha'-dipyridyl,alpha,alpha'-bipyridyl,2,2-bipyridyl PubChem CID: 1474 ChEBI: CHEBI:30351 SMILES: C1=CC=C(N=C1)C1=CC=CC=N1
α,α-Dipyridyl, 99.5%, MP Biomedicals™
CAS: 366-18-7 Molecular Formula: C10H8N2 Molecular Weight (g/mol): 156.19 MDL Number: MFCD00006212 InChI Key: ROFVEXUMMXZLPA-UHFFFAOYSA-N Synonym: 2,2'-bipyridine,2,2'-dipyridyl,2,2'-bipyridyl,bipyridine,2,2'-dipyridine,2-2-pyridyl pyridine,2,2'-bipyridin,alpha,alpha'-dipyridyl,alpha,alpha'-bipyridyl,2,2-bipyridyl PubChem CID: 1474 ChEBI: CHEBI:30351 IUPAC Name: 2,2'-bipyridine SMILES: C1=CC=C(N=C1)C1=CC=CC=N1
2,2'-Bipyridine, ACS, 98%
CAS: 366-18-7 Molecular Formula: C10H8N2 Molecular Weight (g/mol): 156.19 MDL Number: MFCD00006212 InChI Key: ROFVEXUMMXZLPA-UHFFFAOYSA-N Synonym: 2,2'-bipyridine,2,2'-dipyridyl,2,2'-bipyridyl,bipyridine,2,2'-dipyridine,2-2-pyridyl pyridine,2,2'-bipyridin,alpha,alpha'-dipyridyl,alpha,alpha'-bipyridyl,2,2-bipyridyl PubChem CID: 1474 ChEBI: CHEBI:30351 SMILES: C1=CC=C(N=C1)C1=CC=CC=N1
2,2'-Bipyridyl 99.0+%, TCI America™
CAS: 366-18-7 Molecular Formula: C10H8N2 Molecular Weight (g/mol): 156.19 MDL Number: MFCD00006212 InChI Key: ROFVEXUMMXZLPA-UHFFFAOYSA-N Synonym: 2,2'-bipyridine,2,2'-dipyridyl,2,2'-bipyridyl,bipyridine,2,2'-dipyridine,2-2-pyridyl pyridine,2,2'-bipyridin,alpha,alpha'-dipyridyl,alpha,alpha'-bipyridyl,2,2-bipyridyl PubChem CID: 1474 ChEBI: CHEBI:30351 IUPAC Name: 2,2'-bipyridine SMILES: C1=CC=C(N=C1)C1=CC=CC=N1
Sigma Aldrich 2,2'-Bipyridyl
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich 2,2'-Bipyridyl
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Glycolic Acid
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Dimethyl Sulfate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
α,α'-Dipyridyl (Crystalline/Certified), Fisher Chemical
CAS: 366-18-7 Molecular Formula: C10H8N2 Molecular Weight (g/mol): 156.19 MDL Number: MFCD00006212 InChI Key: ROFVEXUMMXZLPA-UHFFFAOYSA-N Synonym: 2,2'-bipyridine,2,2'-dipyridyl,2,2'-bipyridyl,bipyridine,2,2'-dipyridine,2-2-pyridyl pyridine,2,2'-bipyridin,alpha,alpha'-dipyridyl,alpha,alpha'-bipyridyl,2,2-bipyridyl PubChem CID: 1474 ChEBI: CHEBI:30351 IUPAC Name: 2,2'-bipyridine SMILES: C1=CC=C(N=C1)C1=CC=CC=N1
2,2′-Bipyridyl, ≥99.0% (NT), Solstice
CAS: 366-18-7 Molecular Formula: C10H8N2 Molecular Weight (g/mol): 156.19 MDL Number: MFCD00006212 InChI Key: ROFVEXUMMXZLPA-UHFFFAOYSA-N PubChem CID: 1474 ChEBI: CHEBI:30351 IUPAC Name: 2-pyridin-2-ylpyridine SMILES: C1=CC=C(N=C1)C1=CC=CC=N1