Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam 2,2'-Bipyridine, Metalloprotease Inhibitor, >98% Purity, Solid, 10 g

Catalog No. 502704603
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4603 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4603 Supplier Abcam Supplier No. AB14652010G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-affinity metal chelator and metalloprotease inhibitor.

  • Molecular weight: 156.18 g/mol
  • Purity: >98%
  • Form: Solid
  • Function: Metalloprotease inhibitor; high-affinity chelator of iron
  • Notes: Symmetrical bipyridine used as a neutral ligand for chelating with metal ions; may inhibit Fe2+ containing enzymes at 10-8 M

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Ambient
Target Molecule Metal Ions
Form Solid
Buffer 100% 2,2-bipyridine
Content And Storage Store at ambient temperature.
Quantity 10 g
For Use With (Application) Metalloprotease inhibition, Metal ion chelation, Enzyme inhibition
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.