Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2′-Bipyridyl, ≥99.0% (NT), Solstice

Catalog No. 60044862 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-044-862 5 g Glass Bottle
60-044-861 25 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-044-862 Supplier Solstice Supplier No. 144545G

missing translation for 'validMainframeMsg'

Redox indicator, for spectrophotometric det. of Fe,≥99.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 366-18-7
Molecular Formula C10H8N2
Molecular Weight (g/mol) 156.19
MDL Number MFCD00006212
InChI Key ROFVEXUMMXZLPA-UHFFFAOYSA-N
PubChem CID 1474
ChEBI CHEBI:30351
IUPAC Name 2-pyridin-2-ylpyridine
SMILES C1=CC=C(N=C1)C1=CC=CC=N1

Specifications

Melting Point 69°C to 71°C
Boiling Point 273°C
UN Number UN2811
Beilstein 113089
Formula Weight 156.18g/mol
Percent Purity ≥99.0% (NT)
Packaging Glass Bottle
Quantity 5 g
Chemical Name or Material 2,2′-Bipyridyl
Hazard Statement P280-P301 + P310-P312
Precautionary Statement H301 + H311
DOTInformation 6.1/PGIII
EINECSNumber 206-674-4
RTECSNumber DW1750000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.