Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Bis(2-hydroxy-5-biphenylyl)propane 98.0+%, TCI America™
SDP

Catalog No. B27505G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B27505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B27505G Supplier TCI America Supplier No. B27505G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 24038-68-4
Molecular Formula C27H24O2
Molecular Weight (g/mol) 380.487
InChI Key BKTRENAPTCBBFA-UHFFFAOYSA-N
Synonym 5,5′C-Isopropylidenebis(2-hydroxybiphenyl), 4,4′C-Isopropylidenebis(2-phenylphenol)
PubChem CID 13059052
IUPAC Name 4-[2-(4-hydroxy-3-phenylphenyl)propan-2-yl]-2-phenylphenol
SMILES CC(C)(C1=CC(=C(C=C1)O)C2=CC=CC=C2)C3=CC(=C(C=C3)O)C4=CC=CC=C4

Specifications

Melting Point 118°C
Color White
Quantity 5 g
Formula Weight 380.49
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Bis(2-hydroxy-5-biphenylyl)propane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.