Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Bis[4-(4-aminophenoxy)phenyl]hexafluoropropane 98.0+%, TCI America™
SDP

Catalog No. B139825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B139825G 25 g
B13985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B139825G Supplier TCI America Supplier No. B139825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 69563-88-8
Molecular Formula C27H20F6N2O2
Molecular Weight (g/mol) 518.46
MDL Number MFCD00015723
InChI Key HHLMWQDRYZAENA-UHFFFAOYSA-N
PubChem CID 94652
IUPAC Name 4-(4-{2-[4-(4-aminophenoxy)phenyl]-1,1,1,3,3,3-hexafluoropropan-2-yl}phenoxy)aniline
SMILES NC1=CC=C(OC2=CC=C(C=C2)C(C2=CC=C(OC3=CC=C(N)C=C3)C=C2)(C(F)(F)F)C(F)(F)F)C=C1

Specifications

Melting Point 163°C
Color White-Yellow
Quantity 25 g
Synonym 4,4′-(Hexafluoroisopropylidene)bis[(4-aminophenoxy)benzene]
Formula Weight 518.46
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Bis[4-(4-aminophenoxy)phenyl]hexafluoropropane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.