Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Bis(hydroxymethyl)propionic acid, 98%

Catalog No. AC147970050
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC147970050 5 kg Plastic drum
AC147975000 500 g Plastic bottle
AC147970010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC147970050 Supplier Thermo Scientific Chemicals Supplier No. 147970050
Only null left
Add to Cart

CAS: 4767-03-7 | C5H10O4 | 134.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4767-03-7
Molecular Formula C5H10O4
Molecular Weight (g/mol) 134.13
InChI Key PTBDIHRZYDMNKB-UHFFFAOYSA-N

Specifications

Melting Point 185.0°C to 190.0°C
Color White
Flash Point >150°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic drum
Linear Formula CH3C(CH2OH)2CO2H
Quantity 5 kg
Beilstein 03,401
Merck Index 15,3277
Synonym 6,7-dimethoxyquinoxaline-2,3 1h,4h-dione, 2,3-dihydroxy-6,7-dimethoxyquinoxaline, 6,7-dimethoxyquinoxaline-2,3-diol, chembl42430, 6,7-dimethoxy-1,4-dihydro-2,3-quinoxalinedione, acmc-20aogl, 2,3-quinoxalinedione,1,4-dihydro-6,7-dimethoxy, 6,7-dimethoxy-1,4-dihydro-quinoxaline-2,3-dione, 2,3-quinoxalinedione, 1,4-dihydro-6,7-dimethoxy, 6,7-dimethoxy-1,4-dihydro-2,3-quinoxalinedione monohydrate
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,slightly soluble in acetone,insoluble in benzene
SMILES CC(CO)(CO)C(O)=O
IUPAC Name 6,7-dimethoxy-1,4-dihydroquinoxaline-2,3-dione
PubChem CID 613780
Formula Weight 134.13
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2, 2-Bis(hydroxymethyl)propionic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult,
EINECSNumber 225-306-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.