Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Bis(hydroxymethyl)propionic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
500g
1kg
5kg
Packaging:
Plastic bottle
Plastic drum
4767-03-7
C5H10O4
134.13
PTBDIHRZYDMNKB-UHFFFAOYSA-N
2,2-bis hydroxymethyl propionic acid,dimethylolpropionic acid,3-hydroxy-2-hydroxymethyl-2-methylpropanoic acid,2,2-bis hydroxymethyl propanoic acid,propanoic acid, 3-hydroxy-2-hydroxymethyl-2-methyl,2,2-dimethylolpropionic acid,dimethylol propionic acid,2,2-bis hydroxymethyl propanoicacid,unii-4nhi8v17mn,4nhi8v17mn
78501
3-hydroxy-2-(hydroxymethyl)-2-methylpropanoic acid
CC(CO)(CO)C(=O)O
4767-03-7
C5H10O4
134.13
PTBDIHRZYDMNKB-UHFFFAOYSA-N
2,2-bis hydroxymethyl propionic acid,dimethylolpropionic acid,3-hydroxy-2-hydroxymethyl-2-methylpropanoic acid,2,2-bis hydroxymethyl propanoic acid,propanoic acid, 3-hydroxy-2-hydroxymethyl-2-methyl,2,2-dimethylolpropionic acid,dimethylol propionic acid,2,2-bis hydroxymethyl propanoicacid,unii-4nhi8v17mn,4nhi8v17mn
78501
3-hydroxy-2-(hydroxymethyl)-2-methylpropanoic acid
CC(CO)(CO)C(=O)O
Water 0.5% Max. (K.F.)
Packaging Plastic bottle
Quantity 500g
Color White
Melting Point 185.0°C to 190.0°C
Flash Point >150°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula CH3C(CH2OH)2CO2H
Beilstein 03,401
Merck Index 15,3277
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,slightly soluble in acetone,insoluble in benzene
Formula Weight 134.13
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 2, 2-Bis(hydroxymethyl)propionic acid
Show More Show Less

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult,

GHS Signal Word: Warning

EINECSNumber : 225-306-3