Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Bis(trifluoromethyl)benzidine 98.0+%, TCI America™
SDP

Catalog No. B171125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B171125G 25 g
B17111G 1 g
B17115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B171125G Supplier TCI America Supplier No. B171125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 341-58-2
Molecular Formula C14H10F6N2
Molecular Weight (g/mol) 320.24
MDL Number MFCD00190155
InChI Key NVKGJHAQGWCWDI-UHFFFAOYSA-N
Synonym 2,2'-bis trifluoromethyl benzidine, 2,2'-bis trifluoromethyl-1,1'-biphenyl-4,4'-diamine, 4-4-amino-2-trifluoromethyl phenyl-3-trifluoromethyl aniline, 2,2'-bis trifluoromethyl-4,4'-diaminobiphenyl, 2,2'-bis-trifluoromethyl-biphenyl-4,4'-diamine, tfmb, 2,2'-bis trifluoromethyl-4,4'-biphenyldiamine, 4,4'-diamino-2,2'-bis trifluoromethyl biphenyl, 1,1'-biphenyl-4,4'-diamine, 2,2'-bis trifluoromethyl, 4-4-azanyl-2-trifluoromethyl phenyl-3-trifluoromethyl aniline
PubChem CID 629349
IUPAC Name 2,2'-bis(trifluoromethyl)-[1,1'-biphenyl]-4,4'-diamine
SMILES NC1=CC(=C(C=C1)C1=C(C=C(N)C=C1)C(F)(F)F)C(F)(F)F

Specifications

Melting Point 183°C
Quantity 25 g
Formula Weight 320.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Bis(trifluoromethyl)benzidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.