Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-(2-Bromoethyl)-1,3-dioxolane

Catalog No. 23099510G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-099-510G 10 g
23-099-550G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-099-510G Supplier Sigma Aldrich Supplier No. 23099510G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Copper-catalyzed borylation of 2-(2-bromoethyl)-1,3-dioxolane with bis(pinacolato)diboron followed by treatment with potassium bifluoride affords the key organotrifluoroborate reagent.2-(2-bromoethyl)-1,3-dioxolane was used:
  • As starting reagent for the synthesis of (5z,9z)-5,9-hexadecadienoic acid, (5z,9z)-5,9-nonadecadienoic acid and (5z,9z)-5,9-eicosadienoic acid
  • In synthesis of 1-deoxy-castanospermine and 1-deoxy-8a-epi-castanospermine
  • In asymmetric total synthesis of both enantiomers of marine mollusk metabolite puloαupone via evansα asymmetric diels-alder reaction
  • As alkylating agent for amines, dithianes and carboximides
  • With eynamides and sodium azide in a one-pot synthesis of triazoles

Specifications

CAS 18742-02-4
Density 1.542 g/mL (at 25°C (literature))
Boiling Point 68°C to 70°C (8 mmHg)
Molecular Formula C5H9BrO2
Refractive Index n20/D 1.479 (literature)
Linear Formula C5H9BrO2
MDL Number MFCD00003216
Quantity 10 g
Synonym 3-Bromopropionaldehyde ethylene acetal
Molecular Weight (g/mol) 181.03
Percent Purity 96%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.