Learn More
Sigma Organic Chemistry 2- 2-Bromoethyl -1-meth | 1G | MFCD09864560
Supplier: Sigma Organic Chemistry CBR007131G

2- 2-Bromoethyl -1-meth , 1G
About this Item:
MDL #: MFCD09864560
Molecular Weight: 287.04
UNSPSC Code: 12352200
Chemical Formula: C8H17Br2N
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.