Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Butoxyethoxy)ethanol, 99%, Spectrum™ Chemical
SDP

Catalog No. 18608137 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-608-137 4 L Amber Glass Bottle
18-608-134 1 L Amber Glass Bottle
18-608-135 20 L Amber Glass Bottle
18-608-136 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-608-137 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B11784LTGL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 112-34-5
Molecular Formula C8H18O3
Molecular Weight (g/mol) 162.23
InChI Key OAYXUHPQHDHDDZ-UHFFFAOYSA-N
IUPAC Name 2-(2-butoxyethoxy)ethan-1-ol
SMILES CCCCOCCOCCO

Specifications

Acidity 0.0001
CAS Max % 1
Boiling Point 230.4°C (446.7°F)
Packaging Amber Glass Bottle
Quantity 4 L
Percent Purity 0.99
Grade Reagent
Physical Form Liquid
Chemical Name or Material 2-(2-Butoxyethoxy)ethanol

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.