Learn More
Medchemexpress LLC 2-[2-chloro-4-methylsulfonyl-3-(2,2,2-trifluoroethoxymethyl)benzoyl]cyclohexane-1,3-dione | 335104-84-2 | MFCD11840613 | 97.3% | 440.82 g/mol | C17H16ClF3O6S | 5 MG

Supplier: Medchemexpress LLC HY1379105MG
Tembotrione is a triketone herbicide supplied as an analytical standard for research, used to study carotenoid synthesis inhibition and herbicide activity. The material is provided as a small vial with HPLC assay 97.3%, molecular weight 440.82 g/mol, and CAS 335104-84-2. Recommended storage: powder at -20°C (up to 3 years) or 4°C (2 years); in solvent at -80°C (6 months) or -20°C (1 month).
- Triketone herbicide active compound used in research.
- Suitable as an analytical standard for biochemical and analytical assays.
- HPLC purity 97.3% ensures analytical confidence.
- Molecular weight 440.82 g/mol; formula C17H16ClF3O6S.
- Storage conditions specified for powder and solvent forms to preserve stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.