Learn More
Sigma Organic Chemistry 2- 2-Chlorophenoxy acet 1G
Supplier: Sigma Organic Chemistry OTV0007351G

2- 2-Chlorophenoxy acet , 1G
About this item:
MDL #: MFCD02295746
Chemical Formula: C8H6Cl2O2
Molecular Weight: 205.04
UNSPSC Code: 12352200
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.