Learn More
Sigma Organic Chemistry 2-Ethylhexyl 2-cyano-3,3-diphenylacrylate | 250ML | 6197-30-4 | MFCD00059260 | 0.97
Supplier: Sigma Organic Chemistry 415820250ML

2-Ethylhexyl 2-cyano-3,3-diphenylacrylate, 250ML
About This Item:
Synonym(s): 2-Cyano-3,3-diphenylacrylic acid 2-ethylhexyl ester; 2-Ethylhexyl alpha-cyano-beta-phenylcinnamate; 2-Ethylhexyl 2-cyano-3,3-diphenyl-2-propenoate; Octocrylene
Linear Formula: (C6H5)2C=C(CN)CO2CH2CH(C2H5)(CH2)3CH3
Molecular Weight: 361.48
CAS Number: 6197-30-4
MDL Number: MFCD00059260
UNSPSC Code: 12162002
Purity: 0.97
Boiling Point: 218 C/1.5 mmHg (lit.)
Melting Point: -10 C (lit.)
Density: 1.051 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.567 (lit.)
Storage: room temp
EINECS Number: 228-250-8
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.