Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Di-n-octyl-1,3-propanediol 95.0+%, TCI America™
SDP

Catalog No. D28421G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28421G 1 g
D28425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D28421G Supplier TCI America Supplier No. D28421G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 106868-09-1
Molecular Formula C19H40O2
Molecular Weight (g/mol) 300.53
MDL Number MFCD03844790
InChI Key NFPNQEAEXIXGNY-UHFFFAOYSA-N
Synonym 9,9-Bis(hydroxymethyl)heptadecane, 1,3-Dihydroxy-2,2-di-n-octylpropane
PubChem CID 10891950
IUPAC Name 2,2-dioctylpropane-1,3-diol
SMILES CCCCCCCCC(CO)(CO)CCCCCCCC

Specifications

Color White
Quantity 1 g
Formula Weight 300.53
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Di-n-octyl-1,3-propanediol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.