Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Diamino-2'-deoxyadenosine, 98%, Thermo Scientific Chemicals

Catalog No. p-7043145
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ65748MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ65748MD Supplier Thermo Scientific Chemicals Supplier No. J65748MD
Only null left
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 215943-79-6
Molecular Formula C10H15N7O3
Molecular Weight (g/mol) 281.28
MDL Number MFCD09750839
InChI Key DIFJLWYUVQLAHS-SJLGBTOWNA-N
Synonym 2,2'-diamino-2'-deoxyadenosine, 2'-amino-2'-deoxy-2,6-diaminopurineriboside, 2r,3s,4r,5r-4-amino-5-2,6-diamino-9h-purin-9-yl-2-hydroxymethyl tetrahydrofuran-3-ol, 2r,3s,4r,5r-4-amino-5-2,6-diaminopurin-9-yl-2-hydroxymethyl oxolan-3-ol
PubChem CID 71744251
SMILES N[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N1C=NC2=C(N)N=C(N)N=C12

Specifications

Color White
Quantity 250 mg
IUPAC Name (2R,3S,4R,5R)-4-amino-5-(2,6-diamino-9H-purin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
Formula Weight 281.27
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2,2'-Diamino-2'-deoxyadenosine
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.