Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+/-)-2,2'-Dibromo-1,1'-binaphthyl, 96%

Catalog No. AAH2698703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2698703 1 g
AAH2698706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2698703 Supplier Thermo Scientific Chemicals Supplier No. H2698703
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 74866-28-7 | C20H12Br2 | 412.124 g/mol

The synthesis of this class of compounds relies on the lithiation of racemic 2,2'-dibromo-1,1'-binaphthyl with t-BuLi followed by addition of Ph 2 PCl.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The synthesis of this class of compounds relies on the lithiation of racemic 2,2′-dibromo-1,1′-binaphthyl with t-BuLi followed by addition of Ph 2 PCl.

Solubility
Sparingly Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74866-28-7
Molecular Formula C20H12Br2
Molecular Weight (g/mol) 412.124
MDL Number MFCD00188005
InChI Key IJUDEFZBMMRSNM-UHFFFAOYSA-N
Synonym 2,2'-dibromo-1,1'-binaphthyl, 2,2'-dibromo-1,1'-binaphthalene, 1,1'-binaphthalene, 2,2'-dibromo, racemic-2,2'-dibromo-1,1'-binaphthyl, 1r-2,2'-dibromo-1,1'-binaphthalene, 1s-2,2'-dibromo-1,1'-binaphthalene, 2-bromo-1-2-bromonaphthalen-1-yl naphthalene, s---2,2-dibromo-1,1'-binaphthyl, 2,2'-dibromo 1,1' binaphthyl, 1,1'-bi 2-bromonaphthalene
PubChem CID 619909
IUPAC Name 2-bromo-1-(2-bromonaphthalen-1-yl)naphthalene
SMILES C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)Br)Br

Specifications

Boiling Point 178°C to 183°C
Quantity 1 g
Solubility Information Sparingly Soluble in water.
Formula Weight 412.23
Percent Purity 96%
Chemical Name or Material (+/-)-2,2'-Dibromo-1,1'-binaphthyl
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.