Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Dibromobiphenyl, TCI America™
SDP

Catalog No. D28741G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28741G 1 g
D28745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D28741G Supplier TCI America Supplier No. D28741G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 13029-09-9
Molecular Formula C12H8Br2
Molecular Weight (g/mol) 312.004
MDL Number MFCD00093707
InChI Key DRKHIWKXLZCAKP-UHFFFAOYSA-N
Synonym 2,2'-dibromobiphenyl, 2,2'-dibromo-1,1'-biphenyl, o,o'-dibromobiphenyl, 1-bromo-2-2-bromophenyl benzene, 1,1'-biphenyl, 2,2'-dibromo, unii-gsl06p6p2l, biphenyl, 2,2'-dibromo, gsl06p6p2l, 1,1'-biphenyl, 2,2'-dibromo-9ci, biphenyl,2'-dibromo
PubChem CID 83060
IUPAC Name 1-bromo-2-(2-bromophenyl)benzene
SMILES C1=CC=C(C(=C1)C2=CC=CC=C2Br)Br

Specifications

Melting Point 80°C
Color White-Yellow
Quantity 1 g
UN Number 3152
Formula Weight 312.00
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Dibromobiphenyl
EINECSNumber (4)-0018
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.