Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Dichloropropionic Acid 95.0+%, TCI America™
SDP

Catalog No. D133425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D133425G 25 g
D13341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D133425G Supplier TCI America Supplier No. D133425G

Chemical Identifiers

CAS 75-99-0
Molecular Formula C3H4Cl2O2
Molecular Weight (g/mol) 142.963
MDL Number MFCD00004178
InChI Key NDUPDOJHUQKPAG-UHFFFAOYSA-N
Synonym 2,2-dichloropropionic acid, dalapon, kenapon, liropon, proprop, alatex, unipon, sys-omnidel, d-granulat, dawpon-rae
PubChem CID 6418
ChEBI CHEBI:81859
IUPAC Name 2,2-dichloropropanoic acid
SMILES CC(C(=O)O)(Cl)Cl

Specifications

Color Yellow
Boiling Point 99°C
Quantity 25 g
Formula Weight 142.96
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2-Dichloropropionic Acid
EINECSNumber (2)-3913
RTECSNumber UF0690000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.