Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Difluoroacetophenone, 95%

Catalog No. AAH2575003
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2575003 1 g
AAH25750MD 250 mg
AAH2575006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2575003 Supplier Thermo Scientific Chemicals Supplier No. H2575003
Add to Cart

CAS: 395-01-7 | C8H6F2O | 156.132 g/mol

2,2-Difluoroacetophenone acts as an intermediate in the preparation of various substances such as pharmaceuticals. Further, it is used in the one-pot synthesis of difluoromethylated arenes from aryl chlorides and bromides. In addition to this, it undergoes reduction with (-)-diisopinocampheylchloroborane chloride to get R-alcohols of purity 85% ee.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Difluoroacetophenone acts as an intermediate in the preparation of various substances such as pharmaceuticals. Further, it is used in the one-pot synthesis of difluoromethylated arenes from aryl chlorides and bromides. In addition to this, it undergoes reduction with (-)-diisopinocampheylchloroborane chloride to get R-alcohols of purity 85% ee.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 395-01-7
Molecular Formula C8H6F2O
Molecular Weight (g/mol) 156.132
MDL Number MFCD06246879
InChI Key OLYKCPDTXVZOQF-UHFFFAOYSA-N
Synonym 2,2-difluoroacetophenone, alpha,alpha-difluoroacetophenone, 2,2-difluoro-1-phenyl-ethanone, 2,2-difluoro-1-phenylethan-1-one, a,a-difluoroacetophenone, ethanone, 2,2-difluoro-1-phenyl, difluoromethyl phenyl ketone, difluoroacetophenone, acmc-20aojw, acetophenone, 2,2-difluoro
PubChem CID 273286
IUPAC Name 2,2-difluoro-1-phenylethanone
SMILES C1=CC=C(C=C1)C(=O)C(F)F

Specifications

Melting Point 48°C to 50°C
Boiling Point 53°C to 54°C (12 mmHg)
Odor Characteristic
Refractive Index 1.4955
Quantity 1 g
Solubility Information Immiscible with water.
Formula Weight 156.13
Percent Purity 95%
Chemical Name or Material 2,2-Difluoroacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.