Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Difluoroethanol 98.0+%, TCI America™
SDP

Catalog No. D412825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D412825G 25 g
D41285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D412825G Supplier TCI America Supplier No. D412825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 359-13-7
Molecular Formula C2H4F2O
Molecular Weight (g/mol) 82.05
MDL Number MFCD00042248
InChI Key VOGSDFLJZPNWHY-UHFFFAOYSA-N
Synonym 2,2-difluoroethan-1-ol, 2,2-difluoro-ethanol, ethanol, 2,2-difluoro, chf2ch2oh, 2,2-difluroethanol, 2,2-difluorethanol, 2,2-difluoro ethanol, 2,2-difluoroethylalcohol, difluoroethanol related, acmc-209ik0
PubChem CID 136187
IUPAC Name 2,2-difluoroethanol
SMILES C(C(F)F)O

Specifications

Color Colorless
Boiling Point 96°C
Quantity 25 g
UN Number 1987
Formula Weight 82.05
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2-Difluoroethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.