Learn More
Sigma Organic Chemistry 2 2-Difluoroethylamine | 100mg | 430-67-1 | MFCD00798137
Supplier: Sigma Organic Chemistry CDS002768100MG

2 2-Difluoroethylamine , 100mg
About this Item:
CAS #: 430-67-1
MDL #: MFCD00798137
Molecular Weight: 81.06
UNSPSC Code: 12352200
Chemical Formula: C2H5F2N
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.