Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Dimethyl-5-phenyl-1,3-dioxane-4,6-dione 98%, Thermo Scientific™

Catalog No. AC209140500
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC209140500 50 g
AC209142500 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC209140500 Supplier Thermo Scientific Chemicals Supplier No. 209140500

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15231-78-4
Molecular Formula C12H12O4
Molecular Weight (g/mol) 220.224
InChI Key NTUAHNQWHPQAMB-UHFFFAOYSA-N
Synonym 1,3-dioxane-4,6-dione, 2,2-dimethyl-5-phenyl, 5-phenylmeldrum's acid, 5-phenyl-meldrum's acid, acmc-1c8jq, 2.2-dimethyl-5-phenyl-1.3-dioxane-4.6-dione, 1,3-dioxane-4,6-dione,2,2-dimethyl-5-phenyl, 2,2-dimethyl-5-phenyl-1,3 dioxane-4,6-dione
PubChem CID 84848
IUPAC Name 2,2-dimethyl-5-phenyl-1,3-dioxane-4,6-dione
SMILES CC1(OC(=O)C(C(=O)O1)C2=CC=CC=C2)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 50 g
Beilstein 19, IV, 1989
Formula Weight 220.22
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material 2,2-Dimethyl-5-phenyl-1,3-dioxane-4,6-dione, 98%
Show More Show Less
EINECSNumber 239-278-5
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.