Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-[[2-(Dimethylamino)ethyl]methylamino]ethanol 97.0+%, TCI America™
SDP

Catalog No. D4680100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4680100ML 100 mL
D468025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4680100ML Supplier TCI America Supplier No. D4680100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2212-32-0
Molecular Formula C7H18N2O
Molecular Weight (g/mol) 146.23
MDL Number MFCD00010387
InChI Key LSYBWANTZYUTGJ-UHFFFAOYSA-N
PubChem CID 75171
IUPAC Name 2-{[2-(dimethylamino)ethyl](methyl)amino}ethan-1-ol
SMILES CN(C)CCN(C)CCO

Specifications

Color Yellow
Boiling Point 88°C
Quantity 100 mL
Synonym N′-(2-Hydroxyethyl)-N,N,N′-trimethylethylenediamine
Formula Weight 146.23
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-[[2-(Dimethylamino)ethyl]methylamino]ethanol
EINECSNumber (2)-3206
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.