Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Dimethylbutyric acid, 97%

Catalog No. AAA1707218
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1707218 50 g
AAA1707209 10 g
AAA1707230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1707218 Supplier Thermo Scientific Chemicals Supplier No. A1707218
Add to Cart

CAS: 595-37-9 | C6H12O2 | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 595-37-9
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00004200
InChI Key VUAXHMVRKOTJKP-UHFFFAOYSA-N
Synonym 2,2-dimethylbutyric acid, butanoic acid, 2,2-dimethyl, 2,2-dimethyl-butanoic acid, 2,2-dimethyl butyric acid, 2,2-dimethyl butanoic acid, unii-ay606cn05o, butyric acid, 2,2-dimethyl, dimethyl ethyl acetic acid, 2,2-dimethyl-butyric acid, .alpha.,.alpha.-dimethylbutyric acid
PubChem CID 11684
ChEBI CHEBI:38649
IUPAC Name 2,2-dimethylbutanoic acid
SMILES CCC(C)(C)C(=O)O

Specifications

Melting Point -14°C
Density 0.926
Boiling Point 186°C
Flash Point 90°C (194°F)
Odor Pungent
Refractive Index 1.4154
Quantity 50 g
UN Number UN3265
Beilstein 1720666
Formula Weight 116.16
Percent Purity 97%
Chemical Name or Material 2,2-Dimethylbutyric acid
Show More Show Less
Hazard Category H227-H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H314-H318-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 209-865-0
TSCA No

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.