Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Dimethylbutyryl Chloride 98.0+%, TCI America™
SDP

Catalog No. D3987100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3987100G 100 g
D398725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D3987100G Supplier TCI America Supplier No. D3987100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5856-77-9
Molecular Formula C6H11ClO
Molecular Weight (g/mol) 134.60
MDL Number MFCD03093071
InChI Key LDJUYMIFFNTKOI-UHFFFAOYSA-N
Synonym 2,2-dimethylbutyryl chloride, 2,2-dimethyl butyryl chloride, 2,2-dimethyl-butyryl chloride, butanoylchloride, 2,2-dimethyl, 2,2-dimethylbutyric acid chloride, butanoyl chloride, 2,2-dimethyl, 2,2-dimethylbutyrylchloride, acmc-1aznz, ksc274k4n, 2,2-dimethylbutyryl chloride 10g
PubChem CID 79958
IUPAC Name 2,2-dimethylbutanoyl chloride
SMILES CCC(C)(C)C(Cl)=O

Specifications

Color Yellow
Boiling Point 132°C
Quantity 100 g
UN Number 2920
Formula Weight 134.60
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,2-Dimethylbutyryl Chloride
EINECSNumber (2)-0632
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.