Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Dimethylglutaric Anhydride 98.0+%, TCI America™
SDP

Catalog No. D132525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D132525G 25 g
D13255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D132525G Supplier TCI America Supplier No. D132525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2938-48-9
Molecular Formula C7H10O3
Molecular Weight (g/mol) 142.15
MDL Number MFCD00006682
InChI Key PAVNZLVXYJDFNR-UHFFFAOYSA-N
Synonym 2,2-dimethylglutaric anhydride, 2h-pyran-2,6 3h-dione, dihydro-3,3-dimethyl, 3,3-dimethyldihydro-2h-pyran-2,6 3h-dione, dihydro-3,3-dimethyl-2h-pyran-2,6 3h-dione, .alpha.,.alpha.-dimethyl glutaric anhydride, 3,3-dimethyltetrahydro-2h-pyran-2,6-dione, acmc-209h8l, 2,2-dimethylglutaricanhydride, 2,2 dimethylglutaric anhydride, 2.2-dimethylglutaric anhydride
PubChem CID 76247
IUPAC Name 3,3-dimethyloxane-2,6-dione
SMILES CC1(C)CCC(=O)OC1=O

Specifications

Melting Point 38°C
Color White
Boiling Point 270°C
Quantity 25 g
Formula Weight 142.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Dimethylglutaric Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.