Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Dimethylsuccinic Acid 98.0+%, TCI America™
SDP

Catalog No. D218425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D218425G 25 g
D21845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D218425G Supplier TCI America Supplier No. D218425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 597-43-3
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.14
MDL Number MFCD00004196
InChI Key GOHPTLYPQCTZSE-UHFFFAOYSA-N
Synonym 2,2-dimethylsuccinic acid, 2,2-dimethyl-succinic acid, unii-mx6tvr2107, 2,2-dimethyl-butanedioic acid, succinic acid, 2,2-dimethyl, butanedioic acid, 2,2-dimethyl, 2,2-dimethylbutanedioate, 2,2-dimethyl-butanedioicacid, 2,2,-dimethylsuccinate, 2,2-dimethyl succinate
PubChem CID 11701
ChEBI CHEBI:86537
IUPAC Name 2,2-dimethylbutanedioic acid
SMILES CC(C)(CC(O)=O)C(O)=O

Specifications

Melting Point 141°C
Color White
Quantity 25 g
Formula Weight 146.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Dimethylsuccinic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.