Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Dimethylthiazolidine 98.0+%, TCI America™
SDP

Catalog No. D149125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D149125ML 25 mL
D14915ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D149125ML Supplier TCI America Supplier No. D149125ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19351-18-9
Molecular Formula C5H12ClNS
Molecular Weight (g/mol) 153.67
MDL Number MFCD00014488
InChI Key NKPNSVCEUIECCA-UHFFFAOYSA-N
PubChem CID 88015
IUPAC Name hydrogen 2,2-dimethyl-1,3-thiazolidine chloride
SMILES [H+].[Cl-].CC1(C)NCCS1

Specifications

Color Yellow
Boiling Point 61°C
Quantity 25 mL
UN Number 1993
Formula Weight 117.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2-Dimethylthiazolidine
EINECSNumber (5)-5890
RTECSNumber XJ5565700
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.