Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Furyl)benzoic acid, 97%, Thermo Scientific™

Catalog No. CC46201DA
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC46201DA 1 g Amber Glass Bottle
CC46201CB 250 mg Amber Glass Bottle
CC46201DE 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC46201DA Supplier Thermo Scientific Chemicals Supplier No. CC46201DA
Add to Cart

Chemical Identifiers

CAS 331942-47-3
Molecular Formula C11H8O3
Molecular Weight (g/mol) 188.182
InChI Key QRUHYAWZHFTNEA-UHFFFAOYSA-N
Synonym 2-2-furyl benzoic acid, 2-furan-2-yl benzoic acid, 2-fur-2-ylbenzoic acid, benzoic acid,2-2-furanyl, 2-fur-2-yl benzoic acid, 2-furan-2-yl-benzoic acid
PubChem CID 2772293
IUPAC Name 2-(furan-2-yl)benzoic acid
SMILES C1=CC=C(C(=C1)C2=CC=CO2)C(=O)O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 188.18
Percent Purity 97%
Chemical Name or Material 2-(2-Furyl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.