Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Furyl)benzonitrile 97+%, Thermo Scientific™

Catalog No. CC46216CB
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC46216CB 250 mg Amber Glass Bottle
CC46216DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC46216CB Supplier Thermo Scientific Chemicals Supplier No. CC46216CB
Add to Cart

Chemical Identifiers

CAS 155395-45-2
Molecular Formula C11H7NO
Molecular Weight (g/mol) 169.18
MDL Number MFCD04039074
InChI Key XLZNEBADSPRURH-UHFFFAOYSA-N
Synonym 2-2-furyl benzonitrile, 2-furan-2-yl benzonitrile, 2-furan-2-yl-benzonitrile, benzonitrile,2-2-furanyl, 2-2-furanyl benzonitrile, 2-2-furyl benzenecarbonitrile, 2-furan-2-yl benzenecarbonitrile
PubChem CID 4335608
IUPAC Name 2-(furan-2-yl)benzonitrile
SMILES N#CC1=CC=CC=C1C1=CC=CO1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 169.18
Percent Purity 97%
Chemical Name or Material 2-(2-Furyl)benzonitrile
Hazard Category Warning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.