Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Methoxyphenoxy)ethylamine Hydrochloride Hydrate 98.0+%, TCI America™
SDP

Catalog No. M179425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M179425G 25 g
M17945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M179425G Supplier TCI America Supplier No. M179425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 64464-07-9
Molecular Formula C9H14ClNO2
Molecular Weight (g/mol) 203.67
MDL Number MFCD01631120
InChI Key KNWPXZOMSZABHD-UHFFFAOYSA-N
Synonym 2-(2-Aminoethoxy)anisole Hydrochloride, 1-(2-Aminoethoxy)-2-methoxybenzene Hydrochloride
PubChem CID 2762734
IUPAC Name 2-(2-methoxyphenoxy)ethan-1-amine hydrochloride
SMILES Cl.COC1=CC=CC=C1OCCN

Specifications

Color White
Quantity 25 g
Formula Weight 203.67
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(2-Methoxyphenoxy)ethylamine Hydrochloride Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.