Learn More
Sigma Organic Chemistry 2-2-methylpropyl pyrrol | 100mg | MFCD01631843
Supplier: Sigma Organic Chemistry CDS005695100MG

2-2-methylpropyl pyrrol , 100mg
About this Item:
MDL #: MFCD01631843
Molecular Weight: 127.23
UNSPSC Code: 12352200
Chemical Formula: C8H17N
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.