Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Naphthyl)indole, 98%

Catalog No. AAB2043403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2043403 1 g
AAB2043406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2043403 Supplier Thermo Scientific Chemicals Supplier No. B2043403
Add to Cart

CAS: 23746-81-8 | C18H13N | 243.31 g/mol

It is used in the catalytic enantioselective addition of indoles to arylnitroalkenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the catalytic enantioselective addition of indoles to arylnitroalkenes.

Solubility
Soluble in DMSO.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23746-81-8
Molecular Formula C18H13N
Molecular Weight (g/mol) 243.31
MDL Number MFCD00124144
InChI Key CACDYUNTCMPDMI-UHFFFAOYSA-N
Synonym 2-2-naphthyl indole, 2-2-naphthyl-1h-indole, 2-naphthalen-2-yl-1h-indole, acmc-1chvp, 2-2-naphthyl-indol, maybridge1_005904, 2-2-indolyl naphthalene, 2-1h-indol-2-yl naphthalene, 2-2-naphthalenyl-1h-indole
PubChem CID 265232
SMILES N1C2=CC=CC=C2C=C1C1=CC=C2C=CC=CC2=C1

Specifications

Quantity 1 g
Solubility Information Soluble in DMSO.
IUPAC Name 2-naphthalen-2-yl-1H-indole
Formula Weight 243.31
Percent Purity 98%
Chemical Name or Material 2-(2-Naphthyl)indole
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.