Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Sulfonylbis(4-tert-octylphenol) 96.0+%, TCI America™
SDP

Catalog No. S04785G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S04785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S04785G Supplier TCI America Supplier No. S04785G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15452-89-8
Molecular Formula C28H42O4S
Molecular Weight (g/mol) 474.7
MDL Number MFCD00026328
InChI Key LMTGYJHIOQZSAA-UHFFFAOYSA-N
Synonym Bis(2-hydroxy-5-tert-octylphenyl) Sulfone, Bis[2-hydroxy-5-(1,1,3,3-tetramethylbutyl)phenyl] Sulfone
PubChem CID 84923
IUPAC Name 2-[2-hydroxy-5-(2,4,4-trimethylpentan-2-yl)phenyl]sulfonyl-4-(2,4,4-trimethylpentan-2-yl)phenol
SMILES CC(C)(C)CC(C)(C)C1=CC(=C(C=C1)O)S(=O)(=O)C2=C(C=CC(=C2)C(C)(C)CC(C)(C)C)O

Specifications

Melting Point 148°C
Color White-Yellow
Quantity 5 g
Formula Weight 474.70
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Sulfonylbis(4-tert-octylphenol)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.