Learn More
Cayman Chemical 2 2Chlorophenyl2nItrocyc 5mg
SDP
Supplier:  Cayman Chemical 382515 MG
Encompass_Preferred
A DHODH inhibitor (IC50 ~20 nM) selective for DHODH over a panel of greater than 400 kinases at 100 nM induces differentiation of ER-HoxA9 U937 and THP-1 cells in vitro (EC50s ~1 UM) induces differentiation of tumor cells and reduces tumor growth in a THP-1 mouse xenograft model at 15 mg/kg three times daily or 5 mg/kg per day induces differentiation of bone marrow leukemic cells and increases survival In a retroviral transduction mouse model of HoxA9 Meis1 AML
Catalog No. 50-291-6834
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.