Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,3,5-Tetramethylindolenine 98.0+%, TCI America™
SDP

Catalog No. T345825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T345825G 25 g
T34585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T345825G Supplier TCI America Supplier No. T345825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 25981-82-2
Molecular Formula C12H15N
Molecular Weight (g/mol) 173.26
MDL Number MFCD03093020
InChI Key RQVAPBRSUHSDGP-UHFFFAOYSA-N
PubChem CID 117693
IUPAC Name 2,3,3,5-tetramethyl-3H-indole
SMILES CC1=NC2=CC=C(C)C=C2C1(C)C

Specifications

Boiling Point 125°C
Quantity 25 g
Synonym 2,3,3,5-tetramethyl-3h-indole, 2,3,3,5-tetramethylindolenine, 3h-indole, 2,3,3,5-tetramethyl, pubchem2092, 2,3,3,5-tetramethyl-3h-indol, 2,3,3,5-tetramethyl-3-h-indole, 3h-indole,2,3,3,5-tetramethyl, 2,3,3,5-tetramethyl-3-hydroindole, 2,3,3,5-tetramethyl-3-h indole
Formula Weight 173.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3,3,5-Tetramethylindolenine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.