Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,5,6-Pentabromotoluene 98.0+%, TCI America™
SDP

Catalog No. P0827500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0827500G 500 g
P082725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0827500G Supplier TCI America Supplier No. P0827500G

Chemical Identifiers

CAS 87-83-2
Molecular Formula C7H3Br5
Molecular Weight (g/mol) 486.621
MDL Number MFCD00000060
InChI Key OZHJEQVYCBTHJT-UHFFFAOYSA-N
PubChem CID 6906
IUPAC Name 1,2,3,4,5-pentabromo-6-methylbenzene
SMILES CC1=C(C(=C(C(=C1Br)Br)Br)Br)Br

Specifications

Melting Point 290°C
Quantity 500 g
UN Number 3077
Formula Weight 486.62
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,4,5,6-Pentabromotoluene
EINECSNumber (3)-0058
RTECSNumber DA6639500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.