Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',3',4',5',6'-Pentafluoroacetophenone 98.0+%, TCI America™
SDP

Catalog No. P092125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P092125G 25 g
P09215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P092125G Supplier TCI America Supplier No. P092125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 652-29-9
Molecular Formula C8H3F5O
Molecular Weight (g/mol) 210.10
MDL Number MFCD00000296
InChI Key FBGHCYZBCMDEOX-UHFFFAOYSA-N
Synonym 2',3',4',5',6'-pentafluoroacetophenone, 1-perfluorophenyl ethanone, pentafluoroacetophenone, 1-pentafluorophenyl ethan-1-one, ethanone, 1-pentafluorophenyl, 2,3,4,5,6-pentafluoroacetophenone, 1-pentafluorphenyl ethanon, 1-acetyl-2,3,4,5,6-pentafluorobenzene, acmc-209npz, 1-pentafluorophenylethanone
PubChem CID 69546
IUPAC Name 1-(2,3,4,5,6-pentafluorophenyl)ethan-1-one
SMILES CC(=O)C1=C(F)C(F)=C(F)C(F)=C1F

Specifications

Color Yellow
Boiling Point 131°C
Quantity 25 g
Formula Weight 210.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2′,3′,4′,5′,6′-Pentafluoroacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.