Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,5,6-Pentafluorobenzophenone, 98%

Catalog No. AAA1371306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1371306 5 g
AAA1371314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1371306 Supplier Thermo Scientific Chemicals Supplier No. A1371306
Add to Cart

CAS: 1536-23-8 | C13H5F5O | 272.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1536-23-8
Molecular Formula C13H5F5O
Molecular Weight (g/mol) 272.17
MDL Number MFCD00000294
InChI Key HCCPWBWOSASKLG-UHFFFAOYSA-N
Synonym 2,3,4,5,6-pentafluorobenzophenone, pentafluorobenzophenone, perfluorophenyl phenyl methanone, pentafluorophenyl phenyl methanone, methanone, pentafluorophenyl phenyl, benzophenone, 2,3,4,5,6-pentafluoro, 2,3,4,5,6-pentafluorophenyl phenyl methanone, 2,3,4,5,6-pentafluorophenyl phenyl ketone, benzoylpentafluorobenzene, acmc-1bvr7
PubChem CID 73755
IUPAC Name (2,3,4,5,6-pentafluorophenyl)-phenylmethanone
SMILES FC1=C(F)C(F)=C(C(=O)C2=CC=CC=C2)C(F)=C1F

Specifications

Melting Point 36°C to 39°C
Boiling Point 93°C (0.2 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 2059098
Formula Weight 272.18
Percent Purity 98%
Chemical Name or Material 2,3,4,5,6-Pentafluorobenzophenone
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 216-258-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.