Learn More
2,3,4,5,6-Pentafluorophenylacetonitrile, 98%, Thermo Scientific Chemicals
Description
2,3,4,5,6-Pentafluorophenylacetonitrile is used as medicine and liquid crystal materials intermediates. Ortho or para substituted dimers and a mixture of trimers, tetramers and pentamers of PFPA coulb be obtained from 2,3,4,5,6-Pentafluorophenylacetonitrile.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2,3,4,5,6-Pentafluorophenylacetonitrile is used as medicine and liquid crystal materials intermediates. Ortho or para substituted dimers and a mixture of trimers, tetramers and pentamers of PFPA coulb be obtained from 2,3,4,5,6-Pentafluorophenylacetonitrile.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Specifications
Specifications
Density | 1.61 |
Boiling Point | 110°C to 111°C (17 mmHg) |
Flash Point | >110°C (230°F) |
Refractive Index | 1.439 |
Quantity | 5 g |
UN Number | UN3276 |
Beilstein | 1882341 |
Solubility Information | Insoluble in water. |
Formula Weight | 207.1 |
Percent Purity | 98% |
Show More |
Safety and Handling
Hazard Category | H302+H312+H332-H315-H319-H335 |
Hazard Statement | GHS H Statement H331-H302-H312-H315-H319 Toxic if inhaled. Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S. |
EINECSNumber | 211-498-6 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.