Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,5-Tetrafluorobenzeneboronic acid, 97%, Thermo Scientific™

Catalog No. AAL1982403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1982403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1982403 Supplier Thermo Scientific Chemicals Supplier No. L1982403

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 179923-32-1
Molecular Formula C6H3BF4O2
Molecular Weight (g/mol) 193.892
MDL Number MFCD04039308
InChI Key GXGSBMNUHWAXHP-UHFFFAOYSA-N
Synonym 2,3,4,5-tetrafluorophenyl boronic acid, 2,3,4,5-tetrafluorobenzeneboronic acid, tetrafluorophenylboronic acid, acmc-1c04s, 2,3,4,5-tetrafluorophenylboranic acid, boronic acid, 2,3,4,5-tetrafluorophenyl, boronic acid,b-2,3,4,5-tetrafluorophenyl, 2,3,4,5-tetrafluorophenylboronic acid, 2,3,4,5-tetrakis fluoranyl phenyl boronic acid, boronic acid, b-2,3,4,5-tetrafluorophenyl
PubChem CID 2783273
IUPAC Name (2,3,4,5-tetrafluorophenyl)boronic acid
SMILES B(C1=CC(=C(C(=C1F)F)F)F)(O)O

Specifications

Melting Point ∼250°C to 255°C
Quantity 1 g
Beilstein 8554485
Formula Weight 193.9
Percent Purity 97%
Chemical Name or Material 2,3,4,5-Tetrafluorobenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.