Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,5-Tetrafluorobenzonitrile 97.0+%, TCI America™
SDP

Catalog No. T18821G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18821G 1 g
T18825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T18821G Supplier TCI America Supplier No. T18821G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16582-93-7
Molecular Formula C7HF4N
Molecular Weight (g/mol) 175.09
MDL Number MFCD00040187
InChI Key GLTGXIGJLCSEAM-UHFFFAOYSA-N
Synonym 2,3,4,5-tetrafluorobenzenecarbonitrile, tetrafluorobenzonitrile, pubchem9013, intermediates-zcf02243, acmc-1c97m, gltgxigjlcseam-uhfffaoysa, 2,3,4,5-tetrafluorobenzonitrile_x000d_, 2,3,4,5-tetrakis fluoranyl benzenecarbonitrile
PubChem CID 600224
IUPAC Name 2,3,4,5-tetrafluorobenzonitrile
SMILES FC1=CC(C#N)=C(F)C(F)=C1F

Specifications

Color Yellow
Boiling Point 165°C
Quantity 1 g
UN Number 3276
Formula Weight 175.09
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3,4,5-Tetrafluorobenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.