Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,5-Tetrafluorobenzyl Alcohol 98.0+%, TCI America™
SDP

Catalog No. T18831G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18831G 1 g
T18835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T18831G Supplier TCI America Supplier No. T18831G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 53072-18-7
Molecular Formula C7H4F4O
Molecular Weight (g/mol) 180.102
MDL Number MFCD01631452
InChI Key HLUZGUMMQYQHKJ-UHFFFAOYSA-N
Synonym 2,3,4,5-tetrafluorobenzyl alcohol, 2,3,4,5-tetrafluorophenyl methanol, 2,3,4,5-tetrafluorobenzylalcohol, benzenemethanol, 2,3,4,5-tetrafluoro, 2,3,4,5-tetrafluorophenyl methan-1-ol, pubchem4660, benzenemethanol,2,3,4,5-tetrafluoro, 2,3,4,5-tetrafluoro-phenyl-methanol
PubChem CID 2734080
IUPAC Name (2,3,4,5-tetrafluorophenyl)methanol
SMILES C1=C(C(=C(C(=C1F)F)F)F)CO

Specifications

Color Yellow
Quantity 1 g
Formula Weight 180.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3,4,5-Tetrafluorobenzyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.