Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,6-Tetra-O-benzoyl-beta-D-glucopyranosyl Isothiocyanate 98.0+%, TCI America™
SDP

Catalog No. A5515100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5515100MG 100 mg
A55151G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A5515100MG Supplier TCI America Supplier No. A5515100MG

Chemical Identifiers

CAS 132413-50-4
Molecular Formula C35H27NO9S
Molecular Weight (g/mol) 637.66
MDL Number MFCD00075064
InChI Key RITGYQZMPNVVEG-UHFFFAOYNA-N
Synonym BGIT
PubChem CID 11966661
IUPAC Name [3,4,5-tris(benzoyloxy)-6-isothiocyanatooxan-2-yl]methyl benzoate
SMILES O=C(OCC1OC(N=C=S)C(OC(=O)C2=CC=CC=C2)C(OC(=O)C2=CC=CC=C2)C1OC(=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color White-Yellow
Quantity 100 mg
Formula Weight 637.66
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,4,6-Tetra-O-benzoyl-beta-D-glucopyranosyl Isothiocyanate [for HPLC Labeling]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.